Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VMN2R34 SIRNA Assays ›
          Gene Symbol:
          Vmn2r34 Vmn2r28 Vmn2r30 Vmn2r36 Vmn2r37 Vmn2r40 Vmn2r41 Vmn2r45 Vmn2r47 Vmn2r48 Vmn2r52
          Gene Name:
          vomeronasal 2, receptor 34
          vomeronasal 2, receptor 28
          vomeronasal 2, receptor 30
          vomeronasal 2, receptor 36
          vomeronasal 2, receptor 37
          vomeronasal 2, receptor 40
          vomeronasal 2, receptor 41
          vomeronasal 2, receptor 45
          vomeronasal 2, receptor 47
          vomeronasal 2, receptor 48
          vomeronasal 2, receptor 52
          Gene Aliases:
          ENSMUSG00000070841
          EG665255
          ENSMUSG00000070847, V2r15
          100042724, Gm3991
          V2r14
          -
          -
          -
          -
          EG625580
          EG384534
          Chromosome Location:
          Chr.7: 7671829 - 7689398 on Build GRCm38
          Chromosome Location:
          Chr.7: 5480456 - 5493851 on Build GRCm38
          Chromosome Location:
          Chr.7: 7312274 - 7337493 on Build GRCm38
          Chromosome Location:
          Chr.7: 7876653 - 7902462 on Build GRCm38
          Chromosome Location:
          Chr.7: 9205549 - 9223653 on Build GRCm38
          Chromosome Location:
          Chr.7: 8907734 - 8931402 on Build GRCm38
          Chromosome Location:
          Chr.7: 8137905 - 8161551 on Build GRCm38
          Chromosome Location:
          Chr.7: 8471469 - 8488959 on Build GRCm38
          Chromosome Location:
          Chr.7: 9816782 - 9841325 on Build GRCm38
          Chromosome Location:
          Chr.7: 9927624 - 9953585 on Build GRCm38
          Chromosome Location:
          Chr.7: 10158652 - 10176286 on Build GRCm38
          UniGene ID:
          Mm.302158
          Mm.425373
          Mm.458977
          Mm.465202
          Mm.302158
          Mm.469893
          Mm.469892
          Mm.469890
          Mm.465205
          Mm.469893
          Mm.389802
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234566

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Vmn2r34
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105066.1 NP_001098536.1 2 266
          Gene Symbol: Vmn2r28
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081405.1 NP_001074874.1 2 266
          Gene Symbol: Vmn2r30
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009490.3 NP_033516.3 2 266
          GenBank mRNA AF011425.1 - 304
          Gene Symbol: Vmn2r36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105068.1 NP_001098538.1 2 266
          Gene Symbol: Vmn2r37
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009489.2 NP_033515.2 2 381
          GenBank mRNA AF011424.1 - 2 381
          Gene Symbol: Vmn2r40
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105072.1 NP_001098542.1 2 266
          Gene Symbol: Vmn2r41
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105073.1 NP_001098543.1 2 266
          Gene Symbol: Vmn2r45
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105075.1 NP_001098545.1 2 266
          Gene Symbol: Vmn2r47
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105151.1 NP_001098621.1 2 266
          Gene Symbol: Vmn2r48
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105152.1 NP_001098622.1 2 275
          Gene Symbol: Vmn2r52
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105191.1 NP_001098661.1 2 266

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0