Search
Search

Rpl29| Gene Symbol: Rpl29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017150.1 | NP_058846.1 | 4 | 560 | ||
| GenBank mRNA | BC086404.1 | - | 4 | 554 | 7304319 | |
| FQ221016.1 | - | 4 | 561 | |||
| FQ221647.1 | - | 4 | 567 | |||
| FQ229056.1 | - | 4 | 565 | |||
| FQ229057.1 | - | 4 | 565 | |||
| X68283.1 | - | 4 | 560 | |||