Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109610.2 | NP_001103080.1 | 1 | 454 | ||
GenBank mRNA | BC157816.1 | - | 1 | 454 | 9034669 |
Gene Symbol: LOC103690002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763447.1 | XP_008761669.1 | 1 | 493 |