Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109437.2 | NP_001102907.1 | 4 | 557 | ||
GenBank mRNA | BC166797.1 | - | 4 | 557 | 7938214 |