Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC100134871 SIRNA Assays ›
          Gene Symbol:
          LOC100134871 Hbb Hbb-b1 LOC103694855
          Gene Name:
          beta globin minor gene
          hemoglobin, beta
          hemoglobin, beta adult major chain
          hemoglobin subunit beta-2-like
          Gene Aliases:
          -
          -
          -
          -
          Chromosome Location:
          Chr.1: 168957460 - 168958836 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168971269 - 168972680 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 168945529 - 168953023 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906791: 23525 - 31030 on Build Rnor_6.0
          UniGene ID:
          Rn.216394
          Rn.202945
          Rn.36966
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s235212

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC100134871
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113223.1 NP_001106694.1 3 581
          GenBank mRNA FQ209951.1 - 3 584
          FQ210023.1 - 3 584
          FQ210060.1 - 3 582
          FQ210106.1 - 3 582
          FQ210483.1 - 3 582
          FQ210600.1 - 3 584
          FQ210669.1 - 3 578
          FQ211039.1 - 3 576
          FQ211266.1 - 3 582
          FQ211272.1 - 3 582
          FQ211275.1 - 3 582
          FQ211294.1 - 3 584
          FQ211362.1 - 3 582
          FQ211415.1 - 3 582
          FQ211424.1 - 3 582
          FQ211697.1 - 3 584
          FQ212253.1 - 3 582
          FQ212317.1 - 3 584
          FQ212406.1 - 3 584
          FQ212421.1 - 3 582
          FQ213080.1 - 3 611
          FQ214057.1 - 3 582
          FQ214129.1 - 3 582
          FQ217131.1 - 3 580
          FQ217673.1 - 3 582
          FQ218002.1 - 3 583
          FQ218191.1 - 3 582
          FQ218246.1 - 3 554
          FQ218266.1 - 3 620
          FQ218689.1 - 3 582
          FQ218821.1 - 3 582
          FQ219109.1 - 3 582
          FQ219133.1 - 3 613
          FQ221588.1 - 3 580
          FQ222844.1 - 3 582
          FQ224149.1 - 3 582
          FQ224301.1 - 3 582
          FQ224598.1 - 3 582
          FQ224632.1 - 3 582
          FQ224738.1 - 3 582
          FQ224763.1 - 3 621
          FQ225377.1 - 3 582
          FQ225739.1 - 3 584
          FQ226460.1 - 3 582
          FQ226535.1 - 3 582
          FQ226609.1 - 3 584
          FQ228555.1 - 3 582
          FQ230881.1 - 3 584
          FQ232298.1 - 726
          FQ232442.1 - 3 613
          FQ233001.1 - 3 582
          X05080.1 - 3 581
          X16418.1 - 3 547
          Gene Symbol: Hbb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033234.1 NP_150237.1 3 577
          GenBank mRNA BC058448.1 - 3 586 6921704
          FQ209466.1 - 3 582
          FQ209544.1 - 3 582
          FQ209725.1 - 3 584
          FQ209747.1 - 3 582
          FQ209754.1 - 3 582
          FQ209760.1 - 3 582
          FQ209781.1 - 3 582
          FQ209789.1 - 3 582
          FQ209796.1 - 3 582
          FQ209806.1 - 3 584
          FQ209822.1 - 582
          FQ209826.1 - 3 591
          FQ209841.1 - 3 584
          FQ209870.1 - 3 584
          FQ209871.1 - 3 582
          FQ209929.1 - 3 582
          FQ209937.1 - 3 582
          FQ209954.1 - 3 582
          FQ209961.1 - 3 581
          FQ209996.1 - 3 582
          FQ210054.1 - 3 582
          FQ210058.1 - 3 582
          FQ210090.1 - 3 584
          FQ210123.1 - 3 582
          FQ210127.1 - 3 582
          FQ210137.1 - 3 582
          FQ210175.1 - 3 580
          FQ210181.1 - 3 584
          FQ210200.1 - 3 582
          FQ210211.1 - 3 582
          FQ210216.1 - 3 581
          FQ210218.1 - 3 582
          FQ210280.1 - 3 605
          FQ210343.1 - 3 582
          FQ210348.1 - 3 582
          FQ210353.1 - 3 582
          FQ210360.1 - 3 582
          FQ210373.1 - 3 582
          FQ210415.1 - 3 582
          FQ210449.1 - 3 582
          FQ210466.1 - 3 582
          FQ210482.1 - 3 582
          FQ210486.1 - 3 584
          FQ210495.1 - 3 582
          FQ210497.1 - 3 583
          FQ210499.1 - 3 582
          FQ210511.1 - 3 582
          FQ210523.1 - 3 582
          FQ210534.1 - 3 582
          FQ210558.1 - 3 582
          FQ210602.1 - 3 582
          FQ210676.1 - 3 582
          FQ210677.1 - 3 582
          FQ210735.1 - 3 581
          FQ210771.1 - 3 582
          FQ210783.1 - 3 574
          FQ210789.1 - 3 582
          FQ210794.1 - 3 582
          FQ210806.1 - 3 582
          FQ210826.1 - 3 582
          FQ210828.1 - 3 579
          FQ210829.1 - 3 582
          FQ210838.1 - 3 582
          FQ210852.1 - 3 582
          FQ210875.1 - 3 582
          FQ210912.1 - 3 582
          FQ210937.1 - 3 582
          FQ210949.1 - 3 585
          FQ210960.1 - 3 582
          FQ210984.1 - 3 582
          FQ210985.1 - 3 582
          FQ210998.1 - 3 582
          FQ211014.1 - 3 582
          FQ211016.1 - 3 582
          FQ211034.1 - 3 582
          FQ211061.1 - 3 581
          FQ211065.1 - 3 582
          FQ211103.1 - 3 582
          FQ211125.1 - 3 583
          FQ211133.1 - 3 579
          FQ211137.1 - 3 582
          FQ211140.1 - 3 582
          FQ211147.1 - 3 582
          FQ211154.1 - 3 582
          FQ211182.1 - 3 582
          FQ211193.1 - 3 582
          FQ211207.1 - 3 584
          FQ211241.1 - 3 582
          FQ211242.1 - 3 582
          FQ211290.1 - 3 581
          FQ211314.1 - 3 582
          FQ211331.1 - 3 582
          FQ211336.1 - 3 582
          FQ211340.1 - 3 582
          FQ211423.1 - 3 584
          FQ211443.1 - 3 582
          FQ211452.1 - 3 582
          FQ211477.1 - 3 582
          FQ211497.1 - 3 582
          FQ211548.1 - 3 582
          FQ211583.1 - 3 582
          FQ211590.1 - 3 583
          FQ211613.1 - 3 584
          FQ211617.1 - 3 582
          FQ211626.1 - 3 582
          FQ211692.1 - 3 582
          FQ211696.1 - 3 582
          FQ211790.1 - 3 581
          FQ211806.1 - 3 582
          FQ211815.1 - 3 582
          FQ211829.1 - 3 584
          FQ211847.1 - 3 582
          FQ211858.1 - 3 582
          FQ211864.1 - 3 582
          FQ211905.1 - 3 582
          FQ211931.1 - 3 582
          FQ212127.1 - 3 582
          FQ212156.1 - 3 580
          FQ212234.1 - 3 582
          FQ212241.1 - 3 582
          FQ212258.1 - 3 581
          FQ212275.1 - 3 582
          FQ212287.1 - 3 582
          FQ212341.1 - 3 584
          FQ212449.1 - 3 582
          FQ213270.1 - 3 582
          FQ213352.1 - 3 582
          FQ213466.1 - 3 582
          FQ213534.1 - 3 582
          FQ213582.1 - 1061
          FQ213796.1 - 3 582
          FQ214044.1 - 3 582
          FQ214573.1 - 3 582
          FQ216995.1 - 3 584
          FQ217014.1 - 3 584
          FQ217108.1 - 3 582
          FQ217151.1 - 3 582
          FQ217201.1 - 3 582
          FQ217206.1 - 3 582
          FQ217586.1 - 3 584
          FQ217619.1 - 582
          FQ217623.1 - 3 582
          FQ217653.1 - 3 582
          FQ218033.1 - 3 581
          FQ218042.1 - 3 582
          FQ218100.1 - 3 582
          FQ218112.1 - 3 582
          FQ218118.1 - 3 584
          FQ218119.1 - 3 581
          FQ218124.1 - 3 586
          FQ218145.1 - 3 582
          FQ218171.1 - 3 582
          FQ218180.1 - 3 582
          FQ218182.1 - 3 582
          FQ218199.1 - 3 585
          FQ218229.1 - 3 585
          FQ218253.1 - 3 582
          FQ218272.1 - 3 579
          FQ218288.1 - 3 582
          FQ218461.1 - 3 602
          FQ218539.1 - 3 581
          FQ218748.1 - 3 582
          FQ218761.1 - 3 582
          FQ218763.1 - 3 582
          FQ218779.1 - 3 580
          FQ219114.1 - 3 581
          FQ219985.1 - 3 531
          FQ220085.1 - 3 582
          FQ220253.1 - 3 531
          FQ221076.1 - 3 582
          FQ221202.1 - 3 584
          FQ221246.1 - 3 582
          FQ221520.1 - 3 582
          FQ221738.1 - 3 581
          FQ221956.1 - 3 582
          FQ221962.1 - 3 582
          FQ222032.1 - 3 582
          FQ222262.1 - 3 582
          FQ222275.1 - 3 582
          FQ222291.1 - 3 582
          FQ222503.1 - 3 582
          FQ222804.1 - 3 582
          FQ222960.1 - 3 582
          FQ223002.1 - 3 582
          FQ223079.1 - 3 582
          FQ223164.1 - 3 579
          FQ223490.1 - 3 582
          FQ223631.1 - 3 582
          FQ223717.1 - 3 582
          FQ223745.1 - 3 584
          FQ223805.1 - 3 582
          FQ223852.1 - 3 574
          FQ223888.1 - 3 582
          FQ223889.1 - 3 584
          FQ223995.1 - 3 555
          FQ224088.1 - 3 582
          FQ224309.1 - 3 582
          FQ224343.1 - 3 582
          FQ224371.1 - 3 582
          FQ224406.1 - 3 582
          FQ224475.1 - 3 582
          FQ224682.1 - 3 582
          FQ224727.1 - 3 582
          FQ224732.1 - 3 582
          FQ225124.1 - 3 644
          FQ225436.1 - 3 582
          FQ225457.1 - 3 582
          FQ225469.1 - 3 582
          FQ225684.1 - 734
          FQ225738.1 - 3 582
          FQ225754.1 - 3 581
          FQ225816.1 - 3 582
          FQ226188.1 - 3 580
          FQ226536.1 - 3 582
          FQ226572.1 - 3 582
          FQ226889.1 - 3 582
          FQ227120.1 - 3 582
          FQ227498.1 - 3 582
          FQ227822.1 - 3 582
          FQ227875.1 - 3 582
          FQ228016.1 - 3 582
          FQ228244.1 - 3 582
          FQ228281.1 - 3 582
          FQ228294.1 - 3 582
          FQ228295.1 - 3 582
          FQ228296.1 - 3 584
          FQ228414.1 - 3 582
          FQ228616.1 - 3 582
          FQ228638.1 - 3 582
          FQ229015.1 - 3 582
          FQ229033.1 - 3 582
          FQ229489.1 - 3 582
          FQ230471.1 - 3 582
          FQ230486.1 - 3 580
          FQ230503.1 - 3 582
          FQ230508.1 - 3 582
          FQ230541.1 - 3 582
          FQ230861.1 - 3 582
          FQ230866.1 - 3 582
          FQ230889.1 - 3 582
          FQ231191.1 - 3 581
          FQ231208.1 - 3 581
          FQ231253.1 - 3 582
          FQ231286.1 - 3 582
          FQ231289.1 - 3 582
          FQ232655.1 - 3 614
          FQ233311.1 - 3 657
          FQ234713.1 - 3 584
          FQ234942.1 - 3 580
          FQ235254.1 - 1218
          M17084.1 - 3 534
          X16417.1 - 3 577
          Gene Symbol: Hbb-b1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_198776.1 NP_942071.1 3 586
          GenBank mRNA BC058502.1 - 3 586 6887460
          FQ218177.1 - 3 580
          Gene Symbol: LOC103694855
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773799.1 XP_008772021.1 3 622

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell death
          oxygen transport
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          protein heterooligomerization
          oxidation-reduction process
          renal absorption
          platelet aggregation
          glutathione metabolic process
          regulation of eIF2 alpha phosphorylation by heme
          hemopoiesis
          erythrocyte development
          peroxidase activity
          oxygen transporter activity
          iron ion binding
          oxygen binding
          heme binding
          hemoglobin binding
          haptoglobin binding
          hemoglobin alpha binding
          hemoglobin beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0