Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101020.1 | NP_001094490.1 | 1423 | |||
GenBank mRNA | EF501821.1 | - | 1423 |