Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MT1A SIRNA Assays ›
          Gene Symbol:
          Mt1a Ttr
          Gene Name:
          metallothionein 1a
          transthyretin
          Gene Aliases:
          Mt, Mt1
          Lr1, Tbpa
          Chromosome Location:
          Chr.19: 11301991 - 11303007 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 15532963 - 15542180 on Build Rnor_6.0
          UniGene ID:
          Rn.54397
          Rn.1404
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s235497

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mt1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_138826.4 NP_620181.1 3 215
          GenBank mRNA AF411318.1 - 1 49
          BC058442.1 - 3 220 6891030
          DQ255899.1 - 1 17
          FQ209933.1 - 3 216
          FQ210006.1 - 3 216
          FQ210086.1 - 3 216
          FQ210225.1 - 3 216
          FQ210276.1 - 3 216
          FQ210404.1 - 3 216
          FQ210574.1 - 3 216
          FQ210592.1 - 3 216
          FQ210741.1 - 3 216
          FQ210753.1 - 3 216
          FQ210805.1 - 3 216
          FQ210924.1 - 3 216
          FQ210951.1 - 3 216
          FQ211146.1 - 3 216
          FQ211312.1 - 3 216
          FQ211350.1 - 3 216
          FQ217792.1 - 3 216
          FQ218077.1 - 3 216
          FQ218157.1 - 3 216
          FQ218299.1 - 3 216
          FQ218315.1 - 3 216
          FQ218775.1 - 3 216
          FQ220411.1 - 3 216
          FQ221163.1 - 3 216
          FQ221173.1 - 3 216
          FQ221343.1 - 3 216
          FQ221498.1 - 3 216
          FQ221507.1 - 3 216
          FQ221511.1 - 3 216
          FQ221516.1 - 3 216
          FQ221571.1 - 3 216
          FQ221639.1 - 3 216
          FQ221768.1 - 3 216
          FQ221874.1 - 3 216
          FQ222065.1 - 3 216
          FQ222070.1 - 3 216
          FQ222149.1 - 3 216
          FQ222220.1 - 3 216
          FQ222329.1 - 3 216
          FQ222341.1 - 3 216
          FQ222420.1 - 3 216
          FQ222436.1 - 3 216
          FQ222448.1 - 3 216
          FQ222605.1 - 3 216
          FQ222614.1 - 3 215
          FQ222637.1 - 3 216
          FQ222674.1 - 3 216
          FQ222691.1 - 3 216
          FQ222786.1 - 3 216
          FQ222845.1 - 3 216
          FQ222891.1 - 3 216
          FQ222907.1 - 3 216
          FQ222910.1 - 3 216
          FQ222929.1 - 3 216
          FQ222950.1 - 3 216
          FQ222957.1 - 3 215
          FQ223018.1 - 3 216
          FQ223045.1 - 3 216
          FQ223097.1 - 3 216
          FQ223170.1 - 3 216
          FQ223179.1 - 3 216
          FQ223313.1 - 3 216
          FQ223425.1 - 3 216
          FQ223452.1 - 3 216
          FQ228316.1 - 3 216
          FQ228334.1 - 3 216
          FQ228356.1 - 3 216
          FQ228476.1 - 3 216
          FQ228561.1 - 3 216
          FQ228573.1 - 3 216
          FQ228587.1 - 3 216
          FQ228764.1 - 3 216
          FQ228920.1 - 3 216
          FQ228980.1 - 3 216
          FQ229982.1 - 3 216
          J00750.1 - 3 215
          M24327.1 - 1 19
          Gene Symbol: Ttr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA FQ210386.1 - 216

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular metal ion homeostasis
          cellular zinc ion homeostasis
          nitric oxide mediated signal transduction
          response to zinc ion
          detoxification of copper ion
          negative regulation of neuron apoptotic process
          negative regulation of growth
          cellular response to chromate
          cellular response to cadmium ion
          cellular response to zinc ion
          copper ion binding
          zinc ion binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0