Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H2BD SIRNA Assays ›
          Gene Symbol:
          Hist1h2bd Hist1h2bcl1 Hist1h2bf Hist1h2bk Hist1h2bl Hist1h2bo LOC100359692 LOC100365043 LOC100910200
          Gene Name:
          histone cluster 1, H2bd
          histone cluster 1, H2bc-like 1
          histone cluster 1, H2bf
          histone cluster 1, H2bk
          histone cluster 1, H2bl
          histone cluster 1, H2bo
          histone cluster 1, H2bc-like
          histone cluster 1, H2bd-like
          histone H2B type 1-C/E/G-like
          Gene Aliases:
          Hist1h2bm
          Hist1h2bc
          Hist1h2bp
          Hist1h2bc
          H2b
          Hist1h2bm, Hist1h2bn
          -
          -
          -
          Chromosome Location:
          Chr.17: 44833924 - 44834305 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 44737895 - 44738394 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 44748472 - 44748910 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 44527421 - 44527801 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 43817378 - 43817745 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 44756557 - 44758170 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 44519815 - 44520269 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 43808620 - 43809092 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 44792982 - 44793894 on Build Rnor_6.0
          UniGene ID:
          Rn.163361
          -
          -
          Rn.165244
          Rn.112590
          Rn.125401
          -
          -
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s235500

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h2bd
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001108414.1 NP_001101884.1 1 117
          Gene Symbol: Hist1h2bcl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771769.1 XP_008769991.1 1 181
          Gene Symbol: Hist1h2bf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_225384.5 XP_225384.1 1 127
          Gene Symbol: Hist1h2bk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001109400.1 NP_001102870.1 1 117
          Gene Symbol: Hist1h2bl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022647.1 NP_072173.1 1 117
          Gene Symbol: Hist1h2bo
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001106114.1 NP_001099584.1 1 117
          Gene Symbol: LOC100359692
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006254120.2 XP_006254182.1 1 146
          Gene Symbol: LOC100365043
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771682.1 XP_008769904.1 1 170
          Gene Symbol: LOC100910200
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008771772.1 XP_008769994.1 1 175

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001097979, NM_001177653, NM_023422, NM_178201, NM_178196, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          antibacterial humoral response
          defense response to Gram-positive bacterium
          defense response to bacterium
          DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0