Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEDD4L SIRNA Assays ›
Gene Symbol:
NEDD4L
Gene Name:
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases:
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
Chromosome Location:
Chr.18: 58044362 - 58401540 on Build GRCh38
UniGene ID:
Hs.185677
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23571
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEDD4L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144964.1
NP_001138436.1
31
3103
NM_001144965.1
NP_001138437.1
31
3055
NM_001144966.2
NP_001138438.1
30
2980
NM_001144967.2
NP_001138439.1
31
3149
NM_001144968.1
NP_001138440.1
31
3045
NM_001144969.1
NP_001138441.1
30
2985
NM_001144970.2
NP_001138442.1
29
3028
NM_001144971.1
NP_001138443.1
31
3073
NM_001243960.1
NP_001230889.1
29
2957
NM_015277.5
NP_056092.2
30
3089
XM_005266658.4
XP_005266715.2
28
3080
XM_005266660.4
XP_005266717.2
27
3024
XM_005266663.4
XP_005266720.2
26
2964
XM_006722421.3
XP_006722484.2
27
3019
XM_006722424.3
XP_006722487.2
24
2572
XM_006722425.3
XP_006722488.2
23
2516
XM_006722426.3
XP_006722489.1
33
3297
XM_006722428.3
XP_006722491.1
32
3219
XM_006722430.3
XP_006722493.1
31
3127
XM_011525887.2
XP_011524189.1
33
3277
XM_017025676.1
XP_016881165.1
25
2769
XM_017025677.1
XP_016881166.1
24
2713
XM_017025678.1
XP_016881167.1
31
3158
XM_017025679.1
XP_016881168.1
30
3073
XM_017025680.1
XP_016881169.1
27
2761
XM_017025681.1
XP_016881170.1
26
2563
GenBank mRNA
AB007899.1
-
29
2928
AB071179.1
-
30
2920
AF210730.1
-
30
2880
AF385931.1
-
31
3104
AK091189.1
-
3
793
AK130420.1
-
4
1693
AK294963.1
-
30
2980
AK300444.1
-
30
2931
AL137469.1
-
26
2401
AY112983.1
-
31
3055
AY112984.2
-
31
3045
AY112985.2
-
30
2989
AY312514.1
-
31
2866
BC000621.2
-
25
2506
BC019345.1
-
25
2506
BC032597.1
-
29
2984
5528964
BQ215431.1
-
10
713
DQ181796.1
-
31
3073
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
regulation of membrane depolarization
protein monoubiquitination
sodium ion transport
cellular sodium ion homeostasis
excretion
response to metal ion
protein ubiquitination
viral life cycle
water homeostasis
ion transmembrane transport
regulation of ion transmembrane transport
regulation of protein catabolic process
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of protein catabolic process
positive regulation of endocytosis
regulation of membrane repolarization
protein K48-linked ubiquitination
ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
negative regulation of sodium ion transmembrane transport
positive regulation of dendrite extension
negative regulation of protein localization to cell surface
negative regulation of sodium ion transmembrane transporter activity
positive regulation of caveolin-mediated endocytosis
ubiquitin-protein transferase activity
protein binding
potassium channel regulator activity
ligase activity
sodium channel regulator activity
potassium channel inhibitor activity
sodium channel inhibitor activity
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0