Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP1 SIRNA Assays ›
Gene Symbol:
CLASP1
Gene Name:
cytoplasmic linker associated protein 1
Gene Aliases:
MAST1
Chromosome Location:
Chr.2: 121337776 - 121649476 on Build GRCh38
UniGene ID:
Hs.469840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23581
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142273.1
NP_001135745.1
38, 39
4635
NM_001142274.1
NP_001135746.1
37, 38
4611
NM_001207051.1
NP_001193980.1
37, 38
4629
NM_015282.2
NP_056097.1
38, 39
4812
XM_006712377.1
XP_006712440.1
38, 39
4630
XM_006712381.1
XP_006712444.1
36, 37
4579
XM_006712382.1
XP_006712445.1
36, 37
4555
XM_006712383.1
XP_006712446.1
35, 36
4531
XM_006712385.1
XP_006712448.1
31, 32
3699
XM_011510848.1
XP_011509150.1
38, 39
4651
XM_011510849.1
XP_011509151.1
37, 38
4627
XM_017003657.1
XP_016859146.1
42, 43
4933
XM_017003658.1
XP_016859147.1
40, 41
4762
XM_017003659.1
XP_016859148.1
39, 40
4738
XM_017003660.1
XP_016859149.1
39, 40
4732
XM_017003661.1
XP_016859150.1
39, 40
4714
XM_017003662.1
XP_016859151.1
38, 39
4690
XM_017003663.1
XP_016859152.1
38, 39
4684
XM_017003664.1
XP_016859153.1
37, 38
4681
XM_017003665.1
XP_016859154.1
40, 41
4762
XM_017003666.1
XP_016859155.1
39, 40
4654
XM_017003667.1
XP_016859156.1
39, 40
4714
XM_017003668.1
XP_016859157.1
37, 38
4603
XM_017003669.1
XP_016859158.1
38, 39
4690
XM_017003670.1
XP_016859159.1
37, 38
4663
XM_017003671.1
XP_016859160.1
36, 37
4558
XM_017003672.1
XP_016859161.1
37, 38
4603
XM_017003673.1
XP_016859162.1
38, 39
4603
XM_017003674.1
XP_016859163.1
37, 38
4582
XM_017003675.1
XP_016859164.1
36, 37
4558
XM_017003676.1
XP_016859165.1
33, 34
5596
XM_017003677.1
XP_016859166.1
32, 33
5548
XM_017003678.1
XP_016859167.1
31, 32
5523
XM_017003679.1
XP_016859168.1
31, 32
5521
XM_017003680.1
XP_016859169.1
29, 30
5392
XM_017003681.1
XP_016859170.1
31, 32
5439
XM_017003682.1
XP_016859171.1
31, 32
5436
XM_017003683.1
XP_016859172.1
30, 31
5432
XM_017003684.1
XP_016859173.1
32, 33
4529
XM_017003685.1
XP_016859174.1
30, 31
5414
XM_017003686.1
XP_016859175.1
29, 30
5411
XM_017003687.1
XP_016859176.1
31, 32
4505
XM_017003688.1
XP_016859177.1
29, 30
5390
XM_017003689.1
XP_016859178.1
31, 32
4420
GenBank mRNA
AB014522.1
-
30, 31
3675
AF347693.1
-
37, 38
4587
AK074338.1
-
24, 25
2870
AK123216.1
-
5, 6
1922
AK294922.1
-
14, 15
1922
AK297259.1
-
31, 32
3586
AK297420.1
-
7, 8
1134
AK299331.1
-
30, 31
4115
AK301814.1
-
9, 10
1372
AK302393.1
-
31, 32
3716
BC032563.1
-
3, 4
441
BC112940.1
-
37, 38
4555
5501463
BC132723.1
-
38, 39
4597
40146745
BC144107.1
-
37, 38
4591
9052627
BX648933.1
-
20, 21
2461
CR749627.1
-
10, 11
1492
CR933722.1
-
31, 32
3632
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
microtubule cytoskeleton organization
microtubule bundle formation
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
astral microtubule organization
microtubule organizing center organization
negative regulation of microtubule polymerization or depolymerization
positive regulation of microtubule polymerization
microtubule anchoring
establishment of mitotic spindle localization
positive regulation of exocytosis
establishment of spindle orientation
cell division
negative regulation of stress fiber assembly
regulation of focal adhesion assembly
regulation of microtubule cytoskeleton organization
positive regulation of extracellular matrix disassembly
establishment of epithelial cell polarity
mitotic spindle assembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
kinetochore binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0