Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLASP1 SIRNA Assays ›
Gene Symbol:
CLASP1
Gene Name:
cytoplasmic linker associated protein 1
Gene Aliases:
MAST1
Chromosome Location:
Chr.2: 121337776 - 121649476 on Build GRCh38
UniGene ID:
Hs.469840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23582
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLASP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142273.1
NP_001135745.1
26
2797
NM_001142274.1
NP_001135746.1
25
2773
NM_001207051.1
NP_001193980.1
25
2791
NM_015282.2
NP_056097.1
25
2857
XM_006712377.1
XP_006712440.1
26
2792
XM_006712381.1
XP_006712444.1
24
2741
XM_006712382.1
XP_006712445.1
24
2717
XM_006712383.1
XP_006712446.1
23
2693
XM_006712385.1
XP_006712448.1
18
1792
XM_011510848.1
XP_011509150.1
25
2744
XM_011510849.1
XP_011509151.1
24
2720
XM_017003657.1
XP_016859146.1
28
2924
XM_017003658.1
XP_016859147.1
28
2924
XM_017003659.1
XP_016859148.1
27
2900
XM_017003660.1
XP_016859149.1
26
2792
XM_017003661.1
XP_016859150.1
27
2876
XM_017003662.1
XP_016859151.1
26
2852
XM_017003663.1
XP_016859152.1
25
2744
XM_017003664.1
XP_016859153.1
24
2741
XM_017003665.1
XP_016859154.1
28
2924
XM_017003666.1
XP_016859155.1
27
2816
XM_017003667.1
XP_016859156.1
27
2876
XM_017003668.1
XP_016859157.1
25
2765
XM_017003669.1
XP_016859158.1
26
2852
XM_017003670.1
XP_016859159.1
25
2825
XM_017003671.1
XP_016859160.1
24
2720
XM_017003672.1
XP_016859161.1
25
2765
XM_017003673.1
XP_016859162.1
26
2765
XM_017003674.1
XP_016859163.1
25
2744
XM_017003675.1
XP_016859164.1
24
2720
XM_017003676.1
XP_016859165.1
21
3758
XM_017003677.1
XP_016859166.1
20
3710
XM_017003678.1
XP_016859167.1
19
3685
XM_017003679.1
XP_016859168.1
19
3683
XM_017003680.1
XP_016859169.1
17
3554
XM_017003681.1
XP_016859170.1
19
3601
XM_017003682.1
XP_016859171.1
19
3598
XM_017003683.1
XP_016859172.1
18
3594
XM_017003684.1
XP_016859173.1
20
2691
XM_017003685.1
XP_016859174.1
18
3576
XM_017003686.1
XP_016859175.1
17
3573
XM_017003687.1
XP_016859176.1
19
2667
XM_017003688.1
XP_016859177.1
17
3552
XM_017003689.1
XP_016859178.1
19
2582
GenBank mRNA
AB014522.1
-
17
1720
AF347693.1
-
24
2632
AK091140.1
-
24
2731
AK294922.1
-
2
84
AK297259.1
-
19
1748
AK299331.1
-
18
2277
AK302393.1
-
18
1809
BC112940.1
-
25
2717
5501463
BC132723.1
-
26
2759
40146745
BC144107.1
-
25
2753
9052627
BX648933.1
-
7
506
CR933722.1
-
18
1725
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
microtubule cytoskeleton organization
microtubule bundle formation
vesicle targeting
microtubule nucleation
negative regulation of microtubule depolymerization
Golgi organization
mitotic spindle organization
sister chromatid cohesion
establishment or maintenance of cell polarity
exit from mitosis
regulation of gastrulation
positive regulation of epithelial cell migration
regulation of epithelial to mesenchymal transition
astral microtubule organization
microtubule organizing center organization
negative regulation of microtubule polymerization or depolymerization
positive regulation of microtubule polymerization
microtubule anchoring
establishment of mitotic spindle localization
positive regulation of exocytosis
establishment of spindle orientation
cell division
negative regulation of stress fiber assembly
regulation of focal adhesion assembly
regulation of microtubule cytoskeleton organization
positive regulation of extracellular matrix disassembly
establishment of epithelial cell polarity
mitotic spindle assembly
negative regulation of wound healing, spreading of epidermal cells
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
dystroglycan binding
protein binding
microtubule binding
kinetochore binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0