Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SYNE1 SIRNA Assays ›
          Gene Symbol:
          SYNE1
          Gene Name:
          spectrin repeat containing nuclear envelope protein 1
          Gene Aliases:
          8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
          Chromosome Location:
          Chr.6: 152121684 - 152637399 on Build GRCh38
          UniGene ID:
          Hs.12967
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23608

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SYNE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033071.3 NP_149062.1 139 25522
          NM_182961.3 NP_892006.3 139 25831
          XM_006715407.1 XP_006715470.1 140 25813
          XM_006715408.2 XP_006715471.1 140 25805
          XM_006715409.1 XP_006715472.1 139 25792
          XM_006715410.2 XP_006715473.1 140 25817
          XM_006715411.1 XP_006715474.1 139 25762
          XM_006715412.2 XP_006715475.1 140 25802
          XM_006715413.2 XP_006715476.1 139 25748
          XM_006715414.1 XP_006715477.1 138 25741
          XM_006715415.2 XP_006715478.1 139 25748
          XM_006715416.2 XP_006715479.1 139 25733
          XM_006715417.2 XP_006715480.1 139 25676
          XM_006715420.2 XP_006715483.1 139 25664
          XM_006715421.2 XP_006715484.1 139 25661
          XM_006715422.1 XP_006715485.1 139 25654
          XM_006715423.2 XP_006715486.1 140 25817
          XM_006715424.2 XP_006715487.1 140 25817
          XM_006715425.2 XP_006715488.1 139 25748
          XM_011535641.2 XP_011533943.1 140 25814
          XM_011535642.2 XP_011533944.1 140 25802
          XM_011535643.1 XP_011533945.1 139 25648
          XM_011535644.1 XP_011533946.1 126 23864
          XM_011535645.2 XP_011533947.1 122 23257
          XM_017010608.1 XP_016866097.1 141 25932
          XM_017010609.1 XP_016866098.1 140 25739
          XM_017010610.1 XP_016866099.1 139 25718
          XM_017010611.1 XP_016866100.1 139 25521
          XM_017010612.1 XP_016866101.1 138 25445
          XM_017010613.1 XP_016866102.1 139 25745
          XM_017010614.1 XP_016866103.1 139 25661
          XM_017010615.1 XP_016866104.1 138 25592
          XM_017010616.1 XP_016866105.1 139 25748
          XM_017010617.1 XP_016866106.1 139 25745
          XM_017010618.1 XP_016866107.1 139 25733
          XM_017010619.1 XP_016866108.1 126 23896
          GenBank mRNA AB018339.2 - 27 4468
          AF043290.2 - 1 14
          AF444779.1 - 14 2341
          AF495910.1 - 139 25831
          AF535142.1 - 139 25522
          AK122691.1 - 19 3111
          AK127076.1 - 24 3877
          AK304825.1 - 13 2033
          AK316101.1 - 12 2048
          AL833542.1 - 2 4257
          AL833657.1 - 4 584
          AY061755.1 - 54 8921
          AY061756.1 - 12 1980
          AY184203.1 - 11 1845
          AY184206.1 - 52 8816
          BC090927.1 - 12 2093
          BX537837.1 - 21 3409
          BX647837.1 - 25 4072
          JQ754366.1 - 2 374

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          nucleus organization
          Golgi organization
          establishment of nucleus localization
          muscle cell differentiation
          cytoskeletal anchoring at nuclear membrane
          nuclear matrix anchoring at nuclear membrane
          actin binding
          protein binding
          lamin binding
          protein homodimerization activity
          poly(A) RNA binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0