Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DOCK9 SIRNA Assays ›
Gene Symbol:
DOCK9
Gene Name:
dedicator of cytokinesis 9
Gene Aliases:
ZIZ1, ZIZIMIN1
Chromosome Location:
Chr.13: 98793487 - 99086406 on Build GRCh38
UniGene ID:
Hs.596105
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23614
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DOCK9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130048.1
NP_001123520.1
17, 18
2160
NM_001130049.1
NP_001123521.1
17, 18
2043
NM_001130050.1
NP_001123522.1
17, 18
2160
NM_001318849.1
NP_001305778.1
17, 18
2043
NM_015296.2
NP_056111.1
17, 18
2043
XM_005254034.3
XP_005254091.1
17, 18
2032
XM_005254035.3
XP_005254092.1
17, 18
2032
XM_006719922.1
XP_006719985.1
17, 18
2193
XM_006719924.2
XP_006719987.1
17, 18
2032
XM_006719925.1
XP_006719988.1
17, 18
2160
XM_006719931.3
XP_006719994.1
17, 18
1988
XM_006719932.1
XP_006719995.1
17, 18
2160
XM_006719936.1
XP_006719999.1
17, 18
2193
XM_006719937.3
XP_006720000.1
17, 18
1988
XM_017020503.1
XP_016875992.1
17, 18
2033
XM_017020504.1
XP_016875993.1
17, 18
2033
XM_017020505.1
XP_016875994.1
17, 18
2033
XM_017020506.1
XP_016875995.1
17, 18
2033
XM_017020507.1
XP_016875996.1
17, 18
2033
XM_017020508.1
XP_016875997.1
17, 18
2033
XM_017020509.1
XP_016875998.1
17, 18
2160
XM_017020510.1
XP_016875999.1
17, 18
2033
XM_017020511.1
XP_016876000.1
17, 18
2033
XM_017020512.1
XP_016876001.1
17, 18
2160
XM_017020513.1
XP_016876002.1
17, 18
2033
XM_017020514.1
XP_016876003.1
17, 18
2160
XM_017020515.1
XP_016876004.1
17, 18
2160
XM_017020516.1
XP_016876005.1
17, 18
2160
XM_017020517.1
XP_016876006.1
17, 18
2132
XM_017020518.1
XP_016876007.1
17, 18
2132
XM_017020519.1
XP_016876008.1
17, 18
2160
XM_017020520.1
XP_016876009.1
17, 18
2160
XM_017020521.1
XP_016876010.1
17, 18
2193
GenBank mRNA
AB028981.2
-
17, 18
2066
AF527605.1
-
17, 18
2043
AK122987.1
-
6, 7
647
AK126492.1
-
17, 18
2160
AK127329.1
-
17, 18
2132
AK127508.1
-
14, 15
1650
BC136274.1
-
17, 18
2023
9020261
BC146814.1
-
17, 18
2037
8860392
BC171753.1
-
17, 18
2023
9053254
CR936820.1
-
17, 18
2137
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
blood coagulation
biological_process
positive regulation of GTPase activity
cell-cell adhesion
guanyl-nucleotide exchange factor activity
protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0