Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBR4 SIRNA Assays ›
Gene Symbol:
UBR4
Gene Name:
ubiquitin protein ligase E3 component n-recognin 4
Gene Aliases:
RBAF600, ZUBR1, p600
Chromosome Location:
Chr.1: 19074506 - 19210334 on Build GRCh38
UniGene ID:
Hs.148078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23626
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBR4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020765.2
NP_065816.2
74, 75
11072
XM_011541108.2
XP_011539410.1
76, 77
11307
XM_011541109.2
XP_011539411.1
76, 77
11304
XM_011541110.2
XP_011539412.1
76, 77
11304
XM_011541111.2
XP_011539413.1
76, 77
11304
XM_011541112.2
XP_011539414.1
76, 77
11292
XM_011541113.2
XP_011539415.1
76, 77
11289
XM_011541114.2
XP_011539416.1
76, 77
11289
XM_011541115.2
XP_011539417.1
76, 77
11283
XM_011541116.2
XP_011539418.1
75, 76
11274
XM_011541117.2
XP_011539419.1
75, 76
11223
XM_011541118.2
XP_011539420.1
75, 76
11220
XM_011541119.2
XP_011539421.1
76, 77
11307
XM_011541120.2
XP_011539422.1
75, 76
11184
XM_011541121.2
XP_011539423.1
74, 75
11151
XM_017000822.1
XP_016856311.1
76, 77
11286
XM_017000823.1
XP_016856312.1
75, 76
11259
XM_017000824.1
XP_016856313.1
75, 76
11205
XM_017000825.1
XP_016856314.1
74, 75
11190
XM_017000826.1
XP_016856315.1
75, 76
11187
XM_017000827.1
XP_016856316.1
74, 75
11172
XM_017000828.1
XP_016856317.1
74, 75
11148
XM_017000829.1
XP_016856318.1
74, 75
11100
XM_017000830.1
XP_016856319.1
73, 74
11049
GenBank mRNA
AB007931.1
-
15, 16
2348
AF348492.1
-
74, 75
11068
AK022322.1
-
3, 4
597
AK023826.1
-
12, 13
1904
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
viral process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
calmodulin binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline