Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106729.1 | NP_001100199.1 | 3 | 413 | ||
NM_001134987.1 | NP_001128459.1 | 3 | 397 | |||
XM_006240100.2 | XP_006240162.1 | 3 | 438 | |||
GenBank mRNA | BC168880.1 | - | 3 | 397 | 9099474 |