Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198665.1 | NP_001185594.1 | 9 | 958 | ||
NM_001301084.1 | NP_001288013.1 | 10 | 841 | |||
NM_015313.2 | NP_056128.1 | 10 | 1015 | |||
XM_006718805.3 | XP_006718868.1 | 10 | 1374 | |||
XM_011542720.2 | XP_011541022.1 | 10 | 956 | |||
XM_017017421.1 | XP_016872910.1 | 10 | 1231 | |||
GenBank mRNA | AB002380.2 | - | 5 | 377 | ||
AF180681.1 | - | 10 | 687 | |||
AK294803.1 | - | 10 | 816 | |||
AK304316.1 | - | 10 | 1239 | |||
BC063117.1 | - | 9 | 961 | |||
BX647785.1 | - | 10 | 817 | |||
BX648515.1 | - | 10 | 822 |