Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RT1-CE12 SIRNA Assays ›
          Gene Symbol:
          RT1-CE12 RT1-A1 RT1-A2 RT1-A3 RT1-CE1 RT1-CE10 RT1-CE14 RT1-CE2 RT1-CE3 RT1-CE4 RT1-EC2 Rt1.aa
          Gene Name:
          RT1 class I, locus CE12
          RT1 class Ia, locus A1
          RT1 class Ia, locus A2
          RT1 class I, locus A3
          RT1 class I, locus1
          RT1 class I, locus CE10
          RT1 class I, locus CE14
          RT1 class I, locus CE2
          RT1 class I, locus CE3
          RT1 class I, locus CE4
          RT1 class Ib, locus EC2
          MHC class I RT1.Aa alpha-chain
          Gene Aliases:
          -
          RT1.A1(N), RT1A-1
          RT1.A2(N), RT1A-2
          RT1-A, RT1.A3
          RT1-C/E1
          -
          -
          RT1-C/E2
          RT1-C/E3
          RT1-C/E4
          MHCRTA, RATMHCRTA, RT1-A, RT1-Aw2, RT1.Al, RT1.EC2, RT1Aw2, RTS
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.20: 5414464 - 5418012 on Build Rnor_6.0
          Chromosome Location:
          Chr.20: 5375011 - 5378536 on Build Rnor_6.0
          Chromosome Location:
          -
          Chromosome Location:
          Chr.20: 4638235 - 4792390 on Build Rnor_6.0
          Chromosome Location:
          Chr.20: 154072 - 157679 on Build Rnor_6.0
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.20: 4931965 - 4935372 on Build Rnor_6.0
          Chromosome Location:
          Chr.20: 4918523 - 4921339 on Build Rnor_6.0
          Chromosome Location:
          -
          Chromosome Location:
          -
          UniGene ID:
          Rn.128939
          Rn.233151
          Rn.116787
          Rn.116787
          Rn.203332
          Rn.219012
          Rn.234142
          Rn.120283
          Rn.40130
          Rn.215620
          Rn.116785
          Rn.116787
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s236912

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RT1-CE12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008835.1 NP_001008835.1 448
          Gene Symbol: RT1-A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008827.1 NP_001008827.1 3 448
          XM_006256099.2 XP_006256161.1 3 479
          GenBank mRNA FQ228888.1 - 3 495
          FQ231496.1 - 3 468
          KC222922.1 - 2 353
          U50448.1 - 3 439
          X90375.1 - 2 376
          Gene Symbol: RT1-A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008829.1 NP_001008829.1 3 442
          GenBank mRNA BC088465.1 - 3 458 7127909
          FQ233418.1 - 3 468
          X90376.1 - 2 376
          Gene Symbol: RT1-A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008830.1 NP_001008830.1 4 439
          GenBank mRNA U38971.1 - 4 439
          Gene Symbol: RT1-CE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008832.2 NP_001008832.1 3 448
          XM_008772726.1 XP_008770948.1 3 579
          XM_008772727.1 XP_008770949.1 3 582
          GenBank mRNA AJ005022.1 - 2 376
          Gene Symbol: RT1-CE10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008833.1 NP_001008833.1 3 448
          XM_006255847.2 XP_006255909.1 3 453
          GenBank mRNA BC083874.1 - 2 632
          BC098952.1 - 3 451 7366301
          Gene Symbol: RT1-CE14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008837.1 NP_001008837.1 3 448
          NM_001033985.1 NP_001029157.1 2 452
          GenBank mRNA BC083799.1 - 2 452 7108728
          Gene Symbol: RT1-CE2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008840.1 NP_001008840.1 3 439
          Gene Symbol: RT1-CE3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008841.2 NP_001008841.1 3 448
          Gene Symbol: RT1-CE4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008842.1 NP_001008842.1 3 448
          Gene Symbol: RT1-EC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012645.1 NP_036777.1 3 457
          GenBank mRNA AF400159.1 - 3 457
          AJ004887.1 - 2 376
          AJ004888.1 - 2 376
          AJ005024.1 - 2 376
          AJ005025.1 - 2 376
          AJ249698.1 - 3 376
          AJ249699.1 - 2 376
          AJ249700.1 - 2, 3 376
          AJ249701.1 - 3 376
          AJ249702.1 - 3 376
          AJ249703.1 - 3 382
          AJ249704.1 - 2 376
          AJ249705.1 - 2 376
          AJ306619.1 - 3 457
          BC061817.1 - 4 458 5598840
          L26224.1 - 3 457
          L40363.1 - 4 436
          L40365.1 - 3 457
          M24023.1 - 2 226
          M25319.1 - 2 436
          M31018.1 - 4 457
          M31038.1 - 3 457
          U38970.1 - 3 439
          U38972.1 - 4 439
          X82106.1 - 2 376
          X90370.1 - 2 376
          X90371.1 - 3 376
          X90372.1 - 2 376
          X90373.1 - 3 376
          X90374.1 - 2 376
          X99767.1 - 2 376
          Y08532.1 - 3 376
          Y13579.1 - 2 376
          Gene Symbol: Rt1.aa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134701.1 NP_001128173.1 4 453
          GenBank mRNA BC166751.1 - 4 453 5621043
          Y14014.1 - 3 376

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: D87665.
          This siRNA also targets the following transcripts in Mouse: NM_207648, NM_001025208, NM_001143689, NM_010394, NM_010380, and others.

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          antigen processing and presentation of peptide antigen via MHC class I
          T cell tolerance induction
          positive regulation of tolerance induction to nonself antigen
          immune response
          negative regulation of natural killer cell mediated cytotoxicity
          antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
          receptor binding
          protein binding
          beta-2-microglobulin binding
          peptide binding
          peptide antigen binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0