Search
Search

| Gene Symbol: LOC689770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142304.1 | NP_001135776.1 | 256 | |||
| GenBank mRNA | EU128747.1 | - | 262 | |||
| FJ404474.1 | - | 256 | ||||
| Gene Symbol: LOC689757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101019.2 | NP_001094489.2 | 2 | 286 | ||
| XM_008763332.1 | XP_008761554.1 | 2 | 589 | |||
| GenBank mRNA | EF100687.1 | - | 2 | 256 | ||
| FJ404472.1 | - | 2 | 286 | |||