Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323614.1 | NP_001310543.1 | 7 | 890 | ||
NM_001323615.1 | NP_001310544.1 | 7 | 890 | |||
NM_001323616.1 | NP_001310545.1 | 8 | 1022 | |||
NM_001323617.1 | NP_001310546.1 | 8 | 857 | |||
NM_015327.2 | NP_056142.2 | 7 | 829 | |||
XM_017000842.1 | XP_016856331.1 | 3 | 775 | |||
GenBank mRNA | AB029012.2 | - | 7 | 831 | ||
AB085691.1 | - | 7 | 757 | |||
AK092183.1 | - | 7 | 805 | |||
AY168922.1 | - | 7 | 683 | |||
AY336728.1 | - | 7 | 680 | |||
BC038296.1 | - | 7 | 830 | 4824487 |