Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323614.1 | NP_001310543.1 | 2 | 315 | ||
NM_001323615.1 | NP_001310544.1 | 2 | 315 | |||
NM_001323616.1 | NP_001310545.1 | 3 | 447 | |||
NM_001323617.1 | NP_001310546.1 | 3 | 282 | |||
NM_015327.2 | NP_056142.2 | 2 | 254 | |||
XM_017000843.1 | XP_016856332.1 | 2 | 261 | |||
GenBank mRNA | AB029012.2 | - | 2 | 256 | ||
AB085691.1 | - | 2 | 182 | |||
AK092183.1 | - | 2 | 230 | |||
AY168922.1 | - | 2 | 108 | |||
AY336728.1 | - | 2 | 105 | |||
BC038296.1 | - | 2 | 255 | 4824487 | ||
CT000143.1 | - | 2 | 315 |