Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024499.3 | 1 | 1017 | |||
NR_024501.3 | 2 | 635 | ||||
NR_024502.3 | 3 | 475 | ||||
NR_024503.3 | 2 | 371 | ||||
GenBank mRNA | EU251065.1 | - | 1 | 1017 | ||
EU251066.1 | - | 2 | 635 | |||
EU251067.1 | - | 3 | 475 | |||
EU251068.1 | - | 2 | 371 | |||
FJ887036.1 | - | 1 | 978 |