Search
Search

BRD4| Gene Symbol: BRD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014299.2 | NP_055114.1 | 6, 7 | 1429 | ||
| NM_058243.2 | NP_490597.1 | 6, 7 | 1429 | |||
| XM_011527854.1 | XP_011526156.1 | 6, 7 | 1494 | |||
| XM_011527855.1 | XP_011526157.1 | 6, 7 | 1495 | |||
| XM_011527856.2 | XP_011526158.1 | 6, 7 | 1494 | |||
| GenBank mRNA | AF386649.1 | - | 6, 7 | 1429 | ||
| AK307664.1 | - | 6, 7 | 1319 | |||
| BC000156.1 | - | 6, 7 | 1275 | |||
| BC030158.2 | - | 6, 7 | 1296 | |||
| BC035266.1 | - | 6, 7 | 1500 | 4837491 | ||
| BC038988.1 | - | 6, 7 | 1470 | |||
| BC047888.1 | - | 6, 7 | 1483 | |||
| BC067129.1 | - | 6, 7 | 1451 | |||
| BC091649.1 | - | 6, 7 | 1467 | |||
| Y12059.1 | - | 6, 7 | 1429 | |||