Search
Search

SRRM2| Gene Symbol: SRRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016333.3 | NP_057417.3 | 11 | 5631 | ||
| XM_005255226.1 | XP_005255283.1 | 11 | 5609 | |||
| GenBank mRNA | AB002322.3 | - | 11 | 5283 | ||
| AB016091.1 | - | 1 | 653 | |||
| AB016092.1 | - | 11 | 5307 | |||
| AF201422.1 | - | 11 | 5311 | |||
| BX647088.1 | - | 10 | 5177 | |||