Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2400
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
20, 21
1845
NM_001190442.1
NP_001177371.1
21, 22
1717
NM_001284212.2
NP_001271141.1
18, 19
1496
NM_001284213.2
NP_001271142.1
17, 18
1403
NM_173060.3
NP_775083.1
19, 20
1553
NR_104285.1
7, 8
709
XM_006714696.3
XP_006714759.1
23, 24
1859
XM_006714697.3
XP_006714760.1
22, 23
1876
XM_006714698.3
XP_006714761.1
21, 22
1837
XM_006714699.3
XP_006714762.1
22, 23
1800
XM_006714700.3
XP_006714763.1
21, 22
1819
XM_006714701.3
XP_006714764.1
21, 22
1835
XM_006714702.3
XP_006714765.1
21, 22
1711
XM_006714703.3
XP_006714766.1
20, 21
1796
XM_006714704.3
XP_006714767.1
21, 22
1744
XM_006714705.3
XP_006714768.1
20, 21
1753
XM_006714706.3
XP_006714769.1
20, 21
1671
XM_006714707.3
XP_006714770.1
20, 21
1622
XM_006714708.3
XP_006714771.1
19, 20
1582
XM_006714709.3
XP_006714772.1
19, 20
1545
XM_006714710.3
XP_006714773.1
18, 19
1500
XM_006714711.3
XP_006714774.1
18, 19
1545
XM_006714712.3
XP_006714775.1
17, 18
1492
XM_006714713.3
XP_006714776.1
7, 8
709
XM_006714714.3
XP_006714777.1
7, 8
709
XM_006714715.3
XP_006714778.1
8, 9
1435
XM_011543654.2
XP_011541956.1
21, 22
1795
XM_011543655.2
XP_011541957.1
21, 22
1792
XM_011543656.2
XP_011541958.1
20, 21
1739
XM_011543657.2
XP_011541959.1
20, 21
1621
XM_011543658.2
XP_011541960.1
18, 19
1474
XM_017009911.1
XP_016865400.1
21, 22
1837
XM_017009912.1
XP_016865401.1
22, 23
1848
XM_017009913.1
XP_016865402.1
22, 23
1817
XM_017009914.1
XP_016865403.1
21, 22
1819
XM_017009915.1
XP_016865404.1
22, 23
1821
XM_017009916.1
XP_016865405.1
21, 22
1835
XM_017009917.1
XP_016865406.1
21, 22
1753
XM_017009918.1
XP_016865407.1
20, 21
1780
XM_017009919.1
XP_016865408.1
20, 21
1753
XM_017009920.1
XP_016865409.1
20, 21
1663
XM_017009921.1
XP_016865410.1
20, 21
1680
XM_017009922.1
XP_016865411.1
19, 20
1714
XM_017009923.1
XP_016865412.1
19, 20
1714
XM_017009924.1
XP_016865413.1
19, 20
1696
XM_017009925.1
XP_016865414.1
19, 20
1632
XM_017009926.1
XP_016865415.1
19, 20
1632
XM_017009927.1
XP_016865416.1
20, 21
1621
XM_017009928.1
XP_016865417.1
19, 20
1620
XM_017009929.1
XP_016865418.1
19, 20
1539
XM_017009930.1
XP_016865419.1
18, 19
1506
XM_017009931.1
XP_016865420.1
18, 19
1500
XM_017009932.1
XP_016865421.1
17, 18
1563
GenBank mRNA
AB208816.1
-
21, 22
1681
AF327443.1
-
16, 17
1395
AK292289.1
-
7, 8
709
AK292356.1
-
7, 8
682
AK296919.1
-
21, 22
1675
AK297879.1
-
19, 20
1495
AK298541.1
-
20, 21
1622
AK299384.1
-
18, 19
1253
AK299402.1
-
20, 21
1602
AK300624.1
-
17, 18
1386
AK302206.1
-
8, 9
709
AK311585.1
-
8, 9
887
AK311598.1
-
8, 9
763
AK314290.1
-
18, 19
1498
AK315146.1
-
18, 19
1497
AK315673.1
-
7, 8
657
AL832174.1
-
19, 20
1538
AL832349.1
-
19, 20
1541
AY189741.1
-
9, 10
708
BC013579.1
-
20, 21
1548
3878564
BC078149.1
-
15, 16
1131
BT009783.1
-
17, 18
1330
BX648397.1
-
18, 19
1447
D16217.1
-
20, 21
1615
U31345.1
-
9, 10
725
U38525.1
-
2, 3
181
U58996.2
-
7, 8
688
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-wfnwd:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0