Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASP2 SIRNA Assays ›
          Gene Symbol:
          CASP2
          Gene Name:
          caspase 2
          Gene Aliases:
          CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57
          Chromosome Location:
          Chr.7: 143288215 - 143307696 on Build GRCh38
          UniGene ID:
          Hs.368982
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2410
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 562.00
          Your Price
          Online offer: 533.65
          533.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CASP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001224.4 NP_001215.1 2 151
          NM_032982.3 NP_116764.2 2 397
          NM_032983.3 NP_116765.2 2 397
          GenBank mRNA AF314174.1 - 2 105
          AF314175.1 - 1 63
          AK291274.1 - 2 189
          AK303391.1 - 2 169
          BC002427.2 - 2 169 3346279
          BG397039.1 - 2 228
          BT007240.1 - 2 105
          CB988674.1 - 2 330
          CR541748.1 - 1 63
          DA505883.1 - 2 398
          DB455837.1 - 2 362
          DB455840.1 - 2 362
          KU177957.1 - 2 156
          KU177958.1 - 2 156
          U13021.1 - 2 118
          U13022.1 - 2 151

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00154242_m1

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          luteolysis
          neural retina development
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          brain development
          aging
          intrinsic apoptotic signaling pathway in response to DNA damage
          protein processing
          ectopic germ cell programmed cell death
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of neuron apoptotic process
          cellular response to mechanical stimulus
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          execution phase of apoptosis
          positive regulation of apoptotic signaling pathway
          cysteine-type endopeptidase activity
          protein binding
          enzyme binding
          protein domain specific binding
          cysteine-type endopeptidase activity involved in apoptotic process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0