Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMUG1 SIRNA Assays ›
          Gene Symbol:
          SMUG1
          Gene Name:
          single-strand-selective monofunctional uracil-DNA glycosylase 1
          Gene Aliases:
          FDG, HMUDG, UNG3
          Chromosome Location:
          Chr.12: 54171815 - 54188994 on Build GRCh38
          UniGene ID:
          Hs.632721
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s24135

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMUG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243787.1 NP_001230716.1 4 512
          NM_001243788.1 NP_001230717.1 3 428
          NM_001243789.1 NP_001230718.1 5 789
          NM_001243790.1 NP_001230719.1 4 708
          NM_001243791.1 NP_001230720.1 3 428
          NM_014311.2 NP_055126.1 4 515
          NR_045039.1 4 510
          XM_006719319.3 XP_006719382.1 5 614
          XM_006719320.3 XP_006719383.1 4 721
          XM_006719321.3 XP_006719384.1 3 466
          XM_006719322.3 XP_006719385.1 4 514
          XM_011538109.2 XP_011536411.1 5 785
          XM_011538110.2 XP_011536412.1 5 788
          XM_011538111.2 XP_011536413.1 4 689
          XM_011538112.2 XP_011536414.1 4 691
          XM_011538113.2 XP_011536415.1 4 554
          XM_011538114.2 XP_011536416.1 4 710
          XM_011538115.2 XP_011536417.1 4 584
          XM_011538116.2 XP_011536418.1 4 524
          XM_011538117.2 XP_011536419.1 4 500
          XM_011538118.2 XP_011536420.1 4 545
          XM_011538119.2 XP_011536421.1 4 487
          XM_011538120.2 XP_011536422.1 3 465
          XM_011538121.2 XP_011536423.1 5 789
          XM_011538122.2 XP_011536424.1 5 785
          XM_011538123.2 XP_011536425.1 5 785
          XM_017019113.1 XP_016874602.1 5 779
          XM_017019114.1 XP_016874603.1 3 862
          XM_017019115.1 XP_016874604.1 4 587
          XM_017019116.1 XP_016874605.1 6 1041
          XM_017019117.1 XP_016874606.1 4 551
          XM_017019118.1 XP_016874607.1 4 488
          XM_017019119.1 XP_016874608.1 4 525
          XM_017019120.1 XP_016874609.1 4 718
          GenBank mRNA AF125182.1 - 2 397
          AK001235.1 - 4 494
          AK001789.1 - 3 1519
          AK222473.1 - 4 491
          AK290274.1 - 3 420
          BC000417.2 - 5 735 2961516
          BC088352.1 - 4 556 6470294
          BC105607.1 - 3 408 6724466
          BG706635.1 - 3 431
          BG764502.1 - 5 598
          BU195802.1 - 2 480
          CD245413.1 - 2 392
          DA430734.1 - 3 420
          KU178578.1 - 2 365
          KU178579.1 - 2 365

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          base-excision repair
          depyrimidination
          oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
          DNA binding
          uracil DNA N-glycosylase activity
          protein binding
          single-strand selective uracil DNA N-glycosylase activity
          DNA N-glycosylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0