Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMUG1 SIRNA Assays ›
Gene Symbol:
SMUG1
Gene Name:
single-strand-selective monofunctional uracil-DNA glycosylase 1
Gene Aliases:
FDG, HMUDG, UNG3
Chromosome Location:
Chr.12: 54171815 - 54188994 on Build GRCh38
UniGene ID:
Hs.632721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24136
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMUG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243787.1
NP_001230716.1
3
352
NM_001243788.1
NP_001230717.1
2
268
NM_001243789.1
NP_001230718.1
4
629
NM_001243790.1
NP_001230719.1
3
548
NM_001243791.1
NP_001230720.1
2
268
NM_014311.2
NP_055126.1
3
355
NR_045039.1
3
350
XM_006719319.3
XP_006719382.1
4
454
XM_006719320.3
XP_006719383.1
3
561
XM_006719321.3
XP_006719384.1
2
306
XM_006719322.3
XP_006719385.1
3
354
XM_011538109.2
XP_011536411.1
4
625
XM_011538110.2
XP_011536412.1
4
628
XM_011538111.2
XP_011536413.1
3
529
XM_011538112.2
XP_011536414.1
3
531
XM_011538113.2
XP_011536415.1
3
394
XM_011538114.2
XP_011536416.1
3
550
XM_011538115.2
XP_011536417.1
3
424
XM_011538116.2
XP_011536418.1
3
364
XM_011538117.2
XP_011536419.1
3
340
XM_011538118.2
XP_011536420.1
3
385
XM_011538119.2
XP_011536421.1
3
327
XM_011538120.2
XP_011536422.1
2
305
XM_011538121.2
XP_011536423.1
4
629
XM_011538122.2
XP_011536424.1
4
625
XM_011538123.2
XP_011536425.1
4
625
XM_017019113.1
XP_016874602.1
4
619
XM_017019114.1
XP_016874603.1
2
702
XM_017019115.1
XP_016874604.1
3
427
XM_017019116.1
XP_016874605.1
5
881
XM_017019117.1
XP_016874606.1
3
391
XM_017019118.1
XP_016874607.1
3
328
XM_017019119.1
XP_016874608.1
3
365
XM_017019120.1
XP_016874609.1
3
558
GenBank mRNA
AF125182.1
-
1
237
AK001235.1
-
3
334
AK001789.1
-
3
328
AK290274.1
-
2
260
BC000417.2
-
4
575
2961516
BC088352.1
-
3
396
6470294
BC105607.1
-
2
248
6724466
BG706635.1
-
2
271
BG764502.1
-
4
439
BU195802.1
-
1
320
CD245413.1
-
1
232
DA430734.1
-
2
260
KU178578.1
-
1
205
KU178579.1
-
1
205
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
DNA glycosylase
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
base-excision repair
depyrimidination
oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
DNA binding
uracil DNA N-glycosylase activity
protein binding
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0