Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022337.2 | NP_071732.1 | 3 | 626 | ||
GenBank mRNA | AF136172.1 | - | 3 | 547 | ||
AF235022.1 | - | 3 | 590 | |||
AL546159.3 | - | 3 | 625 | |||
BC015808.1 | - | 3 | 592 | 3916084 | ||
BT009842.1 | - | 3 | 543 |