Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBP2 SIRNA Assays ›
Gene Symbol:
OSBP2
Gene Name:
oxysterol binding protein 2
Gene Aliases:
HLM, ORP-4, ORP4, OSBPL1, OSBPL4
Chromosome Location:
Chr.22: 30693782 - 30907824 on Build GRCh38
UniGene ID:
Hs.517546
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24367
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282738.1
NP_001269667.1
13
2031
NM_001282739.1
NP_001269668.1
12
2351
NM_001282740.1
NP_001269669.1
11
1534
NM_001282741.1
NP_001269670.1
11
1591
NM_001282742.1
NP_001269671.1
10
1262
NM_030758.3
NP_110385.1
12
2354
XM_005261465.4
XP_005261522.1
11
2133
XM_006724203.2
XP_006724266.2
11
2130
XM_006724207.2
XP_006724270.1
11
1537
XM_006724208.3
XP_006724271.1
10
1322
XM_011530057.1
XP_011528359.1
12
2250
XM_011530058.1
XP_011528360.1
12
2247
XM_011530060.1
XP_011528362.1
11
1591
XM_011530061.1
XP_011528363.1
10
1235
XM_011530062.1
XP_011528364.1
12
1636
XM_017028708.1
XP_016884197.1
11
2133
XM_017028710.1
XP_016884199.1
12
1743
XM_017028711.1
XP_016884200.1
10
1894
XM_017028712.1
XP_016884201.1
11
1538
GenBank mRNA
AB051451.2
-
12
2262
AF288741.1
-
12
2248
AF323731.1
-
12
2354
AK093196.1
-
11
1534
AK131374.1
-
12
1881
AK294007.1
-
11
2139
AK296354.1
-
11
1591
AK296525.1
-
13
2033
AK300323.1
-
10
1262
BC118914.2
-
12
2268
40037933
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
spermatid development
protein binding
cholesterol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sjvkz:80/100.66.74.89:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0