Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013281.3 | NP_037413.1 | 2 | 1661 | ||
NM_198391.2 | NP_938205.1 | 3 | 1855 | |||
XM_005260682.4 | XP_005260739.1 | 3 | 2373 | |||
XM_011529204.2 | XP_011527506.1 | 4 | 2510 | |||
XM_011529205.2 | XP_011527507.1 | 4 | 2172 | |||
GenBank mRNA | AB040902.1 | - | 2 | 1549 | ||
AF169677.1 | - | 2 | 1558 | |||
AK027297.1 | - | 2 | 1572 | |||
AK027670.1 | - | 1 | 769 | |||
AK027694.1 | - | 1 | 1290 | |||
AK074883.1 | - | 2 | 1602 | |||
AK074909.1 | - | 2 | 1530 | |||
AY358319.1 | - | 1 | 1395 | |||
BC020870.1 | - | 2 | 1568 | 4606852 |