Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282157.1 | NP_001269086.1 | 7 | 818 | ||
NM_012280.3 | NP_036412.1 | 11 | 1240 | |||
NM_177439.2 | NP_803188.1 | 12 | 1340 | |||
XM_005272595.1 | XP_005272652.1 | 11 | 1189 | |||
XM_011543893.2 | XP_011542195.1 | 11 | 1195 | |||
XM_011543894.2 | XP_011542196.1 | 12 | 1191 | |||
XM_011543895.1 | XP_011542197.1 | 11 | 1189 | |||
XM_017029382.1 | XP_016884871.1 | 12 | 1185 | |||
GenBank mRNA | AF063015.1 | - | 10 | 827 | ||
AJ005892.1 | - | 11 | 1077 | |||
AK024023.1 | - | 12 | 1200 | |||
AK297486.1 | - | 7 | 642 | |||
AK315138.1 | - | 11 | 1022 | |||
BC021047.1 | - | 11 | 1008 | |||
BC023584.1 | - | 11 | 1006 | 3619796 | ||
KU178580.1 | - | 10 | 777 | |||
KU178581.1 | - | 10 | 777 |