Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TP53TG3B SIRNA Assays ›
          Gene Symbol:
          TP53TG3B LOC102723655 LOC102723713 TP53TG3 TP53TG3C TP53TG3D TP53TG3E TP53TG3F
          Gene Name:
          TP53 target 3B
          TP53-target gene 3 protein
          TP53-target gene 3 protein
          TP53 target 3
          TP53 target 3C
          TP53 target 3D
          TP53 target 3 family member E
          TP53 target 3 family member F
          Gene Aliases:
          TP53TG3E, TP53TG3F
          -
          -
          P53TG3, TP53TG3A, TP53TG3E, TP53TG3F
          TP53TG3, TP53TG3E, TP53TG3F
          TP53TG3, TP53TG3B, TP53TG3C, TP53TG3E, TP53TG3F
          -
          -
          Chromosome Location:
          Chr.16: 33358385 - 33363478 on Build GRCh38
          Chromosome Location:
          Chr.16|NT_187383: 14735 - 19833 on Build GRCh38
          Chromosome Location:
          Chr.16|NT_187383: 933861 - 936780 on Build GRCh38
          Chromosome Location:
          Chr.16: 32673528 - 32678621 on Build GRCh38
          Chromosome Location:
          Chr.16: 33191770 - 33196858 on Build GRCh38
          Chromosome Location:
          Chr.16: 32251926 - 32255928 on Build GRCh38
          Chromosome Location:
          Chr.16: 33301849 - 33306938 on Build GRCh38
          Chromosome Location:
          Chr.16: 33457155 - 33462249 on Build GRCh38
          UniGene ID:
          Hs.592038
          -
          -
          Hs.592038
          Hs.592038
          Hs.513537
          -
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s24440

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TP53TG3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099687.3 NP_001093157.1 2 836
          NR_110897.1 4 1292
          NR_110911.1 3 1125
          XM_006721084.1 XP_006721147.1 3 977
          XM_011545933.1 XP_011544235.1 3 969
          XM_017023638.1 XP_016879127.1 3 3609
          XM_017023639.1 XP_016879128.1 4 1265
          Gene Symbol: LOC102723655
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006724905.3 XP_006724968.1 2 3329
          XM_017030123.1 XP_016885612.1 3 3617
          XM_017030124.1 XP_016885613.1 3 3609
          Gene Symbol: LOC102723713
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011546237.1 XP_011544539.1 3 1314
          XM_011546238.1 XP_011544540.1 3 1306
          XM_017030126.1 XP_016885615.1 4 1604
          XM_017030127.1 XP_016885616.1 4 1596
          Gene Symbol: TP53TG3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016212.4 NP_057296.1 2 836
          NR_110886.1 4 1292
          NR_110910.1 3 1125
          XM_006721026.1 XP_006721089.1 3 977
          XM_011545779.1 XP_011544081.1 3 969
          XM_017023102.1 XP_016878591.1 3 3609
          XM_017023103.1 XP_016878592.1 4 1265
          GenBank mRNA AB023508.1 - 4 1290
          AK097435.1 - 1 1527
          AL133588.1 - 2 1439
          Gene Symbol: TP53TG3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001205259.2 NP_001192188.1 2 834
          NR_110914.1 3 1123
          XM_006721074.2 XP_006721137.1 3 977
          XM_011545920.1 XP_011544222.1 3 969
          XM_017023591.1 XP_016879080.1 3 3609
          XM_017023592.1 XP_016879081.1 4 1273
          XM_017023593.1 XP_016879082.1 4 1265
          Gene Symbol: TP53TG3D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243722.2 NP_001230651.1 2 832
          NR_110898.1 3 998
          XM_017023633.1 XP_016879122.1 3 2519
          XM_017023634.1 XP_016879123.1 3 2511
          XM_017023635.1 XP_016879124.1 4 2687
          XM_017023636.1 XP_016879125.1 3 2397
          XM_017023637.1 XP_016879126.1 3 2389
          GenBank mRNA AB023509.1 - 2 834
          Gene Symbol: TP53TG3E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006721117.2 XP_006721180.1 3 977
          XM_011546032.1 XP_011544334.1 3 969
          XM_017023950.1 XP_016879439.1 3 3617
          XM_017023951.1 XP_016879440.1 3 3609
          XM_017023952.1 XP_016879441.1 4 1273
          XM_017023953.1 XP_016879442.1 4 1265
          XM_017023954.1 XP_016879443.1 2 3329
          Gene Symbol: TP53TG3F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006721118.1 XP_006721181.1 3 977
          XM_011546033.1 XP_011544335.1 3 969
          XM_017023955.1 XP_016879444.1 3 3617
          XM_017023956.1 XP_016879445.1 3 3609
          XM_017023957.1 XP_016879446.1 4 1273
          XM_017023958.1 XP_016879447.1 4 1265
          XM_017023959.1 XP_016879448.1 2 3329

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0