Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NIPBL SIRNA Assays ›
          Gene Symbol:
          NIPBL
          Gene Name:
          NIPBL, cohesin loading factor
          Gene Aliases:
          CDLS, CDLS1, IDN3, IDN3-B, Scc2
          Chromosome Location:
          Chr.5: 36876759 - 37065819 on Build GRCh38
          UniGene ID:
          Hs.481927
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s24589

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NIPBL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015384.4 NP_056199.2 26 5658
          NM_133433.3 NP_597677.2 26 5658
          XM_005248280.3 XP_005248337.1 26 5647
          XM_005248282.4 XP_005248339.3 20 5149
          XM_006714467.2 XP_006714530.1 26 5647
          XM_006714468.2 XP_006714531.1 25 5449
          XM_011514015.1 XP_011512317.1 26 5647
          XM_017009329.1 XP_016864818.1 26 5647
          XM_017009330.1 XP_016864819.1 18 7921
          XM_017009331.1 XP_016864820.1 25 4021
          GenBank mRNA AJ627032.1 - 26 5627
          AJ640137.1 - 26 5627
          KJ807791.1 - 12 1445

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          metanephros development
          heart morphogenesis
          outflow tract morphogenesis
          cellular response to DNA damage stimulus
          mitotic sister chromatid cohesion
          brain development
          sensory perception of sound
          stem cell population maintenance
          positive regulation of histone deacetylation
          maintenance of mitotic sister chromatid cohesion
          cellular protein localization
          embryonic forelimb morphogenesis
          forelimb morphogenesis
          external genitalia morphogenesis
          positive regulation of multicellular organism growth
          ear morphogenesis
          regulation of hair cycle
          fat cell differentiation
          positive regulation of ossification
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of embryonic development
          embryonic digestive tract morphogenesis
          developmental growth
          eye morphogenesis
          regulation of developmental growth
          embryonic viscerocranium morphogenesis
          cognition
          face morphogenesis
          gall bladder development
          uterus morphogenesis
          cellular response to X-ray
          chromatin binding
          protein binding
          protein C-terminus binding
          mediator complex binding
          histone deacetylase binding
          protein N-terminus binding
          chromo shadow domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0