Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKD2 SIRNA Assays ›
          Gene Symbol:
          PRKD2
          Gene Name:
          protein kinase D2
          Gene Aliases:
          HSPC187, PKD2, nPKC-D2
          Chromosome Location:
          Chr.19: 46674316 - 46717127 on Build GRCh38
          UniGene ID:
          Hs.466987
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s24644
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001079880.1 NP_001073349.1 17 2693
          NM_001079881.1 NP_001073350.1 17 2557
          NM_001079882.1 NP_001073351.1 15 2294
          NM_016457.4 NP_057541.2 16 2972
          XM_005258716.2 XP_005258773.2 15 1950
          GenBank mRNA AF309082.1 - 16 2254
          AK074673.1 - 15 1933
          AK075266.1 - 15 2043
          AK092896.1 - 16 2344
          AK095884.1 - 17 2236
          AK300339.1 - 17 2507
          AK303629.1 - 15 1948
          AL050147.1 - 10 1209
          BC015472.1 - 7 671
          BC025307.1 - 12 1543

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00212828_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          positive regulation of endothelial cell proliferation
          adaptive immune response
          protein phosphorylation
          cell adhesion
          cell death
          positive regulation of endothelial cell migration
          peptidyl-serine phosphorylation
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of interleukin-2 production
          positive regulation of interleukin-8 production
          positive regulation of CREB transcription factor activity
          positive regulation of peptidyl-serine phosphorylation
          intracellular signal transduction
          cellular response to vascular endothelial growth factor stimulus
          positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
          positive regulation of blood vessel endothelial cell migration
          positive regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          T cell receptor signaling pathway
          positive regulation of T cell receptor signaling pathway
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          endothelial tube morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          protein kinase D signaling
          mitophagy in response to mitochondrial depolarization
          positive regulation of histone deacetylase activity
          positive regulation of intracellular signal transduction
          positive regulation of DNA biosynthetic process
          positive regulation of endothelial cell chemotaxis
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          protein binding
          ATP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0