Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI3BP SIRNA Assays ›
Gene Symbol:
ABI3BP
Gene Name:
ABI family member 3 binding protein
Gene Aliases:
NESHBP, TARSH
Chromosome Location:
Chr.3: 100749335 - 100993508 on Build GRCh38
UniGene ID:
Hs.477015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24682
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI3BP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015429.3
NP_056244.2
5
722
XM_005247281.2
XP_005247338.1
5
725
XM_005247282.2
XP_005247339.1
5
725
XM_005247283.2
XP_005247340.1
5
725
XM_005247284.2
XP_005247341.1
5
725
XM_005247285.2
XP_005247342.1
5
725
XM_005247287.2
XP_005247344.1
5
725
XM_005247288.2
XP_005247345.1
5
725
XM_005247289.2
XP_005247346.1
5
725
XM_005247290.2
XP_005247347.1
5
725
XM_005247291.2
XP_005247348.1
5
725
XM_005247293.2
XP_005247350.1
5
725
XM_005247294.2
XP_005247351.1
5
725
XM_005247295.2
XP_005247352.1
5
725
XM_005247296.2
XP_005247353.1
5
725
XM_005247298.2
XP_005247355.1
5
725
XM_005247299.3
XP_005247356.1
5
740
XM_005247300.2
XP_005247357.1
5
725
XM_005247301.2
XP_005247358.1
5
725
XM_005247302.2
XP_005247359.1
5
725
XM_005247303.2
XP_005247360.1
5
725
XM_005247305.2
XP_005247362.1
5
725
XM_005247306.2
XP_005247363.1
5
725
XM_005247309.1
XP_005247366.1
3
358
XM_006713569.2
XP_006713632.1
5
725
XM_006713570.2
XP_006713633.1
5
725
XM_006713571.2
XP_006713634.1
5
725
XM_006713572.2
XP_006713635.1
5
725
XM_011512639.1
XP_011510941.1
5
725
XM_011512640.1
XP_011510942.1
5
725
XM_011512641.1
XP_011510943.1
5
725
XM_011512643.1
XP_011510945.1
5
725
XM_011512644.1
XP_011510946.1
5
725
XM_011512645.1
XP_011510947.1
5
725
XM_011512646.1
XP_011510948.1
5
725
XM_011512647.1
XP_011510949.1
5
725
XM_011512648.1
XP_011510950.1
5
725
XM_011512649.1
XP_011510951.1
5
725
XM_011512651.1
XP_011510953.1
5
725
XM_011512652.1
XP_011510954.1
5
725
XM_011512653.1
XP_011510955.1
5
725
XM_011512654.1
XP_011510956.1
5
725
XM_011512656.1
XP_011510958.1
5
725
XM_011512657.1
XP_011510959.1
5
725
XM_011512658.1
XP_011510960.1
5
725
XM_011512659.1
XP_011510961.1
5
725
XM_011512660.1
XP_011510962.1
5
725
XM_017006105.1
XP_016861594.1
5
725
XM_017006106.1
XP_016861595.1
5
725
XM_017006107.1
XP_016861596.1
5
725
XM_017006108.1
XP_016861597.1
5
725
GenBank mRNA
AK025204.1
-
5
682
AL833204.1
-
5
685
BC030221.1
-
5
702
5205716
BX648726.1
-
4
429
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001014423, NM_178790, NM_001014424, NM_001014399, NM_001014422, and others.
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-substrate adhesion
extracellular matrix organization
collagen binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0