Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042549.1 | NP_001036014.1 | 7 | 879 | ||
NM_001297736.1 | NP_001284665.1 | 4 | 671 | |||
NM_001297737.1 | NP_001284666.1 | 5 | 726 | |||
NM_015471.3 | NP_056286.3 | 6 | 794 | |||
XM_017000968.1 | XP_016856457.1 | 7 | 2290 | |||
XM_017000969.1 | XP_016856458.1 | 7 | 2416 | |||
GenBank mRNA | AF201941.1 | - | 6 | 766 | ||
AK222996.1 | - | 6 | 764 | |||
AK223260.1 | - | 6 | 774 | |||
AK313730.1 | - | 6 | 790 | |||
AL050084.1 | - | 6 | 573 |