Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC31B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015490.3 | NP_056305.1 | 26 | 3595 | ||
GenBank mRNA | AF274863.1 | - | 26 | 3591 | ||
AK093637.1 | - | 3 | 1904 | |||
AK098002.1 | - | 2 | 885 | |||
AL080141.1 | - | 19 | 2703 | |||
BC034946.1 | - | 25 | 3834 | |||
BC044569.1 | - | 25 | 3827 | |||
BC053949.1 | - | 25 | 3827 | |||
BC071580.1 | - | 26 | 4071 | |||
BC144313.1 | - | 25 | 3533 | 9052833 | ||
BX640914.1 | - | 25 | 3180 |