Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004352.3 | NP_004343.1 | 2, 3 | 757 | ||
GenBank mRNA | AK314276.1 | - | 2, 3 | 731 | ||
BC093692.1 | - | 3, 4 | 435 | 7939537 | ||
BC093718.1 | - | 3, 4 | 435 | 7939563 | ||
M58583.1 | - | 2, 3 | 705 |