Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300906.1 | NP_001287835.1 | 23 | 3789 | ||
NM_015525.3 | NP_056340.2 | 23 | 3834 | |||
XM_006715453.2 | XP_006715516.1 | 23 | 4374 | |||
XM_006715454.2 | XP_006715517.1 | 23 | 4329 | |||
GenBank mRNA | AB037838.2 | - | 22 | 3558 | ||
AK126777.1 | - | 7 | 1699 | |||
AL050018.1 | - | 3 | 374 | |||
BC027490.1 | - | 1 | 18 | |||
BC038244.2 | - | 6 | 672 | |||
BC042171.1 | - | 2 | 217 | |||
BC113696.1 | - | 22 | 3363 | 8322748 | ||
BC113698.1 | - | 22 | 3363 | 8322750 | ||
BC143736.1 | - | 21 | 2760 | 9052250 | ||
BC143745.1 | - | 22 | 3319 | 9052259 | ||
BC143748.1 | - | 21 | 2760 | |||
DQ005633.1 | - | 23 | 3834 | |||
DQ005634.1 | - | 23 | 3834 |