Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3MBTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015478.6 | NP_056293.4 | 6 | 868 | ||
NM_032107.4 | NP_115479.4 | 9 | 1039 | |||
GenBank mRNA | AB014581.1 | - | 2 | 118 | ||
AK299199.1 | - | 9 | 1009 | |||
AK310345.1 | - | 6 | 868 | |||
BC039820.1 | - | 6 | 783 | 5199689 |