Search
Search

CCK| Gene Symbol: CCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000729.5 | NP_000720.1 | 5 | 1171 | ||
| NM_001174138.2 | NP_001167609.1 | 3 | 404 | |||
| GenBank mRNA | AK289716.1 | - | 3 | 346 | ||
| BC008283.1 | - | 3 | 404 | 3639109 | ||
| BC093055.1 | - | 3 | 346 | 30722619 | ||
| BC111026.1 | - | 5 | 1146 | 5199880 | ||
| BI756101.1 | - | 3 | 332 | |||
| BT006991.1 | - | 2 | 277 | |||
| CB961869.1 | - | 3 | 373 | |||
| CD250282.1 | - | 3 | 506 | |||
| CR542226.1 | - | 2 | 277 | |||