Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286771.2 | NP_001273700.1 | 22 | 3787 | ||
NM_015574.1 | NP_056389.1 | 22 | 4166 | |||
NM_032217.4 | NP_115593.3 | 22 | 4169 | |||
NM_198889.2 | NP_942592.1 | 21 | 3416 | |||
XM_005265671.3 | XP_005265728.1 | 21 | 3757 | |||
XM_005265672.3 | XP_005265729.1 | 22 | 4513 | |||
XM_005265673.3 | XP_005265730.1 | 22 | 4513 | |||
XM_017008011.1 | XP_016863500.1 | 22 | 3741 | |||
XM_017008012.1 | XP_016863501.1 | 21 | 2991 | |||
XM_017008013.1 | XP_016863502.1 | 21 | 2988 | |||
XM_017008014.1 | XP_016863503.1 | 22 | 4510 | |||
XM_017008015.1 | XP_016863504.1 | 22 | 3744 | |||
XM_017008016.1 | XP_016863505.1 | 21 | 3760 | |||
XM_017008017.1 | XP_016863506.1 | 21 | 2991 | |||
GenBank mRNA | AB014597.2 | - | 22 | 3690 | ||
AK025859.1 | - | 7 | 939 | |||
AK055209.1 | - | 14 | 1644 | |||
AK299046.1 | - | 16 | 2669 | |||
AK300302.1 | - | 21 | 3545 | |||
AK307581.1 | - | 21 | 3416 | |||
AL831903.1 | - | 10 | 1969 | |||
AL833287.1 | - | 18 | 2593 | |||
BC004173.2 | - | 22 | 4150 | |||
BC004891.2 | - | 3 | 422 | |||
BC007747.2 | - | 20 | 3270 | |||
BC029935.2 | - | 5 | 694 | |||
BX648035.1 | - | 22 | 3742 |