Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015585.3 | NP_056400.3 | 16 | 1886 | ||
XM_005260688.3 | XP_005260745.1 | 16 | 1849 | |||
XM_005260689.2 | XP_005260746.1 | 16 | 1849 | |||
XM_005260690.3 | XP_005260747.1 | 15 | 1774 | |||
XM_005260691.3 | XP_005260748.1 | 15 | 1771 | |||
XM_005260692.3 | XP_005260749.1 | 15 | 1771 | |||
XM_005260693.3 | XP_005260750.1 | 16 | 1849 | |||
XM_005260695.4 | XP_005260752.1 | 11 | 2160 | |||
XM_005260696.4 | XP_005260753.1 | 7 | 733 | |||
XM_005260697.1 | XP_005260754.1 | 16 | 1849 | |||
XM_011529211.1 | XP_011527513.1 | 15 | 1707 | |||
XM_011529212.2 | XP_011527514.1 | 16 | 1849 | |||
XM_017027786.1 | XP_016883275.1 | 16 | 1789 | |||
XM_017027787.1 | XP_016883276.1 | 16 | 1789 | |||
XM_017027788.1 | XP_016883277.1 | 15 | 1771 | |||
XM_017027789.1 | XP_016883278.1 | 16 | 1849 | |||
XM_017027790.1 | XP_016883279.1 | 16 | 1849 | |||
XM_017027791.1 | XP_016883280.1 | 6 | 631 | |||
XM_017027792.1 | XP_016883281.1 | 7 | 673 | |||
XM_017027793.1 | XP_016883282.1 | 6 | 655 | |||
XM_017027794.1 | XP_016883283.1 | 6 | 719 | |||
XM_017027795.1 | XP_016883284.1 | 15 | 1771 | |||
GenBank mRNA | AK304174.1 | - | 16 | 1789 | ||
BC028708.2 | - | 16 | 1848 | 4825941 |