Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DCAF4 SIRNA Assays ›
Gene Symbol:
DCAF4
Gene Name:
DDB1 and CUL4 associated factor 4
Gene Aliases:
WDR21, WDR21A
Chromosome Location:
Chr.14: 72926332 - 72962098 on Build GRCh38
UniGene ID:
Hs.331491
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25086
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DCAF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163508.1
NP_001156980.1
12
1348
NM_001163509.1
NP_001156981.1
12
1337
NM_015604.3
NP_056419.2
12
1366
NM_181340.2
NP_851937.1
11
1266
NM_181341.2
NP_851938.2
10
1186
XM_017021201.1
XP_016876690.1
11
1178
XM_017021202.1
XP_016876691.1
11
1178
XM_017021203.1
XP_016876692.1
12
1364
XM_017021204.1
XP_016876693.1
12
1334
XM_017021205.1
XP_016876694.1
13
1503
XM_017021206.1
XP_016876695.1
12
1339
XM_017021207.1
XP_016876696.1
12
1361
XM_017021208.1
XP_016876697.1
12
1361
XM_017021209.1
XP_016876698.1
12
1364
XM_017021210.1
XP_016876699.1
12
1361
XM_017021211.1
XP_016876700.1
12
1308
XM_017021212.1
XP_016876701.1
13
1471
XM_017021213.1
XP_016876702.1
12
1271
XM_017021214.1
XP_016876703.1
12
1317
XM_017021215.1
XP_016876704.1
12
1331
GenBank mRNA
AK027745.1
-
12
1193
AK297742.1
-
12
1180
AK300786.1
-
10
1038
AK315499.1
-
12
1366
AL080157.1
-
11
1137
BC018979.2
-
12
1178
3604168
BC035901.1
-
11
1120
5241663
BX161489.1
-
6
811
BX247962.1
-
12
1193
Back To Top
More Information
Gene Ontology Categories:
Function(s)
protein ubiquitination
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0