Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KRIT1 SIRNA Assays ›
          Gene Symbol:
          KRIT1
          Gene Name:
          KRIT1, ankyrin repeat containing
          Gene Aliases:
          CAM, CCM1
          Chromosome Location:
          Chr.7: 92198969 - 92246128 on Build GRCh38
          UniGene ID:
          Hs.531987
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2510

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KRIT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013406.1 NP_001013424.1 4, 5 657
          NM_004912.3 NP_004903.2 5, 6 1152
          NM_194454.1 NP_919436.1 5, 6 805
          NM_194455.1 NP_919437.1 5, 6 1279
          NM_194456.1 NP_919438.1 6, 7 1044
          XM_005250660.3 XP_005250717.1 5, 6 772
          XM_005250662.3 XP_005250719.1 5, 6 751
          XM_005250665.3 XP_005250722.1 5, 6 679
          XM_005250666.3 XP_005250723.1 6, 7 855
          XM_005250667.2 XP_005250724.1 4, 5 563
          XM_005250668.3 XP_005250725.1 4, 5 625
          XM_005250669.3 XP_005250726.1 4, 5 603
          XM_006716161.3 XP_006716224.1 5, 6 903
          XM_006716162.3 XP_006716225.1 5, 6 720
          XM_006716163.3 XP_006716226.1 4, 5 570
          XM_011516651.2 XP_011514953.1 6, 7 886
          XM_011516653.2 XP_011514955.1 4, 5 640
          XM_011516654.2 XP_011514956.1 6, 7 904
          XM_011516655.2 XP_011514957.1 4, 5 558
          XM_011516656.2 XP_011514958.1 4, 5 527
          XM_011516657.2 XP_011514959.1 4, 5 447
          XM_011516658.2 XP_011514960.1 4, 5 459
          XM_011516659.2 XP_011514961.1 4, 5 481
          XM_011516660.2 XP_011514962.1 3, 4 363
          XM_011516661.2 XP_011514963.1 3, 4 441
          XM_017012755.1 XP_016868244.1 4, 5 530
          XM_017012756.1 XP_016868245.1 4, 5 492
          XM_017012757.1 XP_016868246.1 4, 5 524
          XM_017012758.1 XP_016868247.1 5, 6 1130
          XM_017012759.1 XP_016868248.1 4, 5 982
          GenBank mRNA AF296765.2 - 5, 6 1049
          AF310133.1 - 5, 6 744
          AF388384.1 - 5, 6 1279
          AJ294850.1 - 6, 7 1003
          AK055305.1 - 4, 5 657
          AK056537.1 - 4, 5 587
          AY380057.1 - 5, 6 744
          AY380058.1 - 5, 6 744
          AY380059.1 - 5, 6 744
          AY380060.1 - 5, 6 744
          AY993944.1 - 2, 3 260
          BC094684.1 - 2, 3 270 6174749
          BX648659.1 - 1, 2 125

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of endothelial cell proliferation
          small GTPase mediated signal transduction
          negative regulation of endothelial cell migration
          negative regulation of angiogenesis
          positive regulation of protein binding
          cell redox homeostasis
          regulation of catalytic activity
          regulation of establishment of cell polarity
          negative regulation of endothelial cell apoptotic process
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          microtubule binding
          GTPase regulator activity
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0