Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025930.3 | NP_001021100.3 | 9 | 1549 | ||
GenBank mRNA | AF078842.1 | - | 5 | 655 | ||
AK023960.1 | - | 9 | 1166 | |||
AK097236.1 | - | 3 | 1787 | |||
AK125875.1 | - | 5 | 3617 | |||
AK297289.1 | - | 6 | 893 | |||
AK315140.1 | - | 7 | 939 | |||
AL096725.1 | - | 3 | 1328 | |||
AL833939.1 | - | 5 | 1976 | |||
BC098298.1 | - | 3 | 796 | 40025661 | ||
BC098361.1 | - | 3 | 796 | |||
BC099735.1 | - | 3 | 796 | |||
BC105638.1 | - | 3 | 796 | |||
BM717061.1 | - | 1 | 202 |
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 10 | 1830 | ||
GenBank mRNA | BC025289.1 | - | 9 | 1264 |