Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018959.3 | NP_061832.2 | 5 | 527 | ||
NM_170711.2 | NP_733829.1 | 5 | 527 | |||
XM_005259531.3 | XP_005259588.1 | 5 | 477 | |||
XM_005259534.3 | XP_005259591.1 | 5 | 486 | |||
XM_005259535.3 | XP_005259592.1 | 5 | 485 | |||
XM_005259536.3 | XP_005259593.1 | 5 | 488 | |||
XM_011527904.2 | XP_011526206.1 | 5 | 1792 | |||
XM_011527905.2 | XP_011526207.1 | 5 | 1800 | |||
XM_011527906.2 | XP_011526208.1 | 5 | 1797 | |||
XM_011527907.2 | XP_011526209.1 | 5 | 1806 | |||
XM_011527908.2 | XP_011526210.1 | 5 | 1799 | |||
XM_011527909.2 | XP_011526211.1 | 5 | 1799 | |||
XM_011527910.2 | XP_011526212.1 | 5 | 879 | |||
XM_017026581.1 | XP_016882070.1 | 5 | 522 | |||
XM_017026582.1 | XP_016882071.1 | 5 | 1815 | |||
GenBank mRNA | AK056850.1 | - | 3 | 150 | ||
AK125032.1 | - | 4 | 1823 | |||
BC012062.1 | - | 5 | 395 | 4549444 | ||
BI909277.1 | - | 5 | 511 | |||
BU557183.1 | - | 524 | ||||
KU178593.1 | - | 5 | 322 |