Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014394.2 | NP_055209.2 | 8 | 1075 | ||
GenBank mRNA | AB009685.1 | - | 7 | 871 | ||
AF060923.1 | - | 8 | 949 | |||
AF078863.1 | - | 8 | 1000 | |||
AF131820.1 | - | 8 | 1002 | |||
AK098770.1 | - | 9 | 888 | |||
AK130094.1 | - | 8 | 978 | |||
AK130097.1 | - | 8 | 943 | |||
AK225318.1 | - | 8 | 952 | |||
AK292864.1 | - | 8 | 943 | |||
AK297960.1 | - | 8 | 868 | |||
AK298332.1 | - | 8 | 957 | |||
AK299937.1 | - | 7 | 981 | |||
AL136713.1 | - | 8 | 985 | |||
AY358824.1 | - | 8 | 950 | |||
BC010354.1 | - | 8 | 946 | 4093037 | ||
BC022872.1 | - | 8 | 957 | |||
CR533519.1 | - | 7 | 871 |