Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD4 SIRNA Assays ›
          Gene Symbol:
          CD4
          Gene Name:
          CD4 molecule
          Gene Aliases:
          CD4mut
          Chromosome Location:
          Chr.12: 6789472 - 6820810 on Build GRCh38
          UniGene ID:
          Hs.631659
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2578

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000616.4 NP_000607.1 4 554
          NM_001195014.2 NP_001181943.1 2 273
          NM_001195015.2 NP_001181944.1 2 273
          NM_001195016.2 NP_001181945.1 2 273
          NM_001195017.2 NP_001181946.1 2 273
          XM_017020228.1 XP_016875717.1 2 216
          GenBank mRNA AK300052.1 - 2 191
          AK303968.1 - 1 170
          AK308344.1 - 4 397
          AK310652.1 - 3 325
          AK310687.1 - 3 386
          AK310750.1 - 2 209
          AK311495.1 - 2 209
          AK312828.1 - 4 491
          AK315898.1 - 2 209
          AK316461.1 - 2 217
          AK316462.1 - 2 210
          BC025782.1 - 4 448 5226427
          BT019791.1 - 3 296
          BT019811.1 - 3 296
          DA619688.1 - 2 219
          DA639898.1 - 2 219
          M12807.1 - 3 371

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          adaptive immune response
          induction by virus of host cell-cell fusion
          immune response
          signal transduction
          cell surface receptor signaling pathway
          enzyme linked receptor protein signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of calcium ion transport into cytosol
          fusion of virus membrane with host plasma membrane
          T cell differentiation
          entry into host cell
          T cell costimulation
          response to estradiol
          maintenance of protein location in cell
          response to vitamin D
          helper T cell enhancement of adaptive immune response
          positive regulation of T cell proliferation
          T cell selection
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of protein kinase activity
          regulation of defense response to virus by virus
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-negative bacterium
          positive regulation of calcium-mediated signaling
          T cell receptor signaling pathway
          regulation of T cell activation
          virus receptor activity
          glycoprotein binding
          receptor activity
          transmembrane signaling receptor activity
          extracellular matrix structural constituent
          protein binding
          zinc ion binding
          coreceptor activity
          immunoglobulin binding
          enzyme binding
          protein kinase binding
          MHC class II protein binding
          protein homodimerization activity
          protein tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0