Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291501.1 | NP_001278430.1 | 15 | 2415 | ||
NM_148674.4 | NP_683515.4 | 15 | 2415 | |||
XM_011530144.2 | XP_011528446.1 | 15 | 2276 | |||
XM_011530145.2 | XP_011528447.1 | 15 | 2467 | |||
GenBank mRNA | AJ504806.2 | - | 15 | 2415 | ||
AK125736.1 | - | 2 | 174 | |||
BC126208.1 | - | 15 | 2395 | 8991924 |